Methyl 2,3,4-trihydroxybenzoate

Lieferant: Apollo Scientific
Achtung

OR110772-1G
APOSOR110772-1GEA 0 EUR
APOSOR110772-1G
Methyl 2,3,4-trihydroxybenzoate
Methyl 2,3,4-trihydroxybenzoate
Formel: C₈H₈O₅
Molecular Weight: 184,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02149451
CAS-Nummer: 56128-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR