(2,4-Dichlorphenoxy)essigsäurehydrazid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,4-Dichlorphenoxy)acetohydrazid, (2,4-Dichlorphenoxy)acethydrazid

OR11078-25G
APOSOR11078-25GEA 190 EUR
APOSOR11078-25G
(2,4-Dichlorphenoxy)essigsäurehydrazid
(2,4-Dichlorphenoxy)essigsäurehydrazid
Formel: (C₆H₃Cl₂)OCH₂COONHNH₂
Molecular Weight: 235.07 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051972
CAS-Nummer: 28236-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR