5-Methyl-4-nitro-1H-1,2,3-benzotriazole

Lieferant: Apollo Scientific
Achtung

OR110807-500MG
APOSOR110807-500MGEA 48 EUR
APOSOR110807-500MG
5-Methyl-4-nitro-1H-1,2,3-benzotriazole
5-Methyl-4-nitro-1H-1,2,3-benzotriazole
Formel: C₇H₆N₄O₂
Molecular Weight: 178,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00962617
CAS-Nummer: 31995-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR