7-Bromo-1-benzothiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110808-1G
APOSOR110808-1GEA 85.7 EUR
APOSOR110808-1G
7-Bromo-1-benzothiophene-2-carbohydrazide
7-Bromo-1-benzothiophene-2-carbohydrazide
Formel: C₉H₇BrN₂OS
Molecular Weight: 271,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12032269
CAS-Nummer: 1171927-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR