4-(3-Chlorophenyl)-1-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid

Lieferant: Apollo Scientific

OR110813-1G
APOSOR110813-1GEA 429 EUR
APOSOR110813-1G
4-(3-Chlorophenyl)-1-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
4-(3-Chlorophenyl)-1-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
Formel: C₁₀H₈ClN₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18074542
CAS-Nummer: 1355170-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR