Ethyl-8-aminoimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110821-100MG
APOSOR110821-100MGEA 85.7 EUR
APOSOR110821-100MG
Ethyl-8-aminoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-8-aminoimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₀H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11505034
CAS-Nummer: 129912-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR