Imidazo[1,2-a]pyridin-8-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR110823-1G
APOSOR110823-1GEA
24.8
EUR
APOSOR110823-1G
Imidazo[1,2-a]pyridin-8-carbonsäure
Imidazo[1,2-a]pyridin-8-carbonsäure
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,1454 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739251 CAS-Nummer: 133427-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...