3-Amino-3-(4-ethoxyphenyl)propan-1-ol

Lieferant: Apollo Scientific
Achtung

OR110831-1G
APOSOR110831-1GEA 371 EUR
APOSOR110831-1G
3-Amino-3-(4-ethoxyphenyl)propan-1-ol
3-Amino-3-(4-ethoxyphenyl)propan-1-ol
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412722
CAS-Nummer: 683221-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR