Ethyl-2-amino-5-iodbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-amino-5-iodobenzoate

OR11084-5G
APOSOR11084-5GEA 21.9 EUR
APOSOR11084-5G
Ethyl-2-amino-5-iodbenzoat
Ethyl-2-amino-5-iodbenzoat
Formel: C₉H₁₀INO₂
Molecular Weight: 291,09 g/mol
Schmelzpunkt: 70…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038940
CAS-Nummer: 268568-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR