N-(tert-Butyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Lieferant: Apollo Scientific

OR110845-1G
APOSOR110845-1GEA 296 EUR
APOSOR110845-1G
N-(tert-Butyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
N-(tert-Butyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Formel: C₁₀H₁₅N₅
Molecular Weight: 205,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03180509
CAS-Nummer: 869072-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR