tert-Butyl N-(5-methylthiophen-2-yl)carbamate
Lieferant: Apollo Scientific
OR110846-1G
APOSOR110846-1GEA
371
EUR
APOSOR110846-1G
tert-Butyl N-(5-methylthiophen-2-yl)carbamate
tert-Butyl N-(5-methylthiophen-2-yl)carbamate
Formel:
C₁₀H₁₅NO₂S Molecular Weight: 213,297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02940774 CAS-Nummer: 62188-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...