5-Bromoindan-1-ol

Lieferant: Apollo Scientific

OR110851-1G
APOSOR110851-1GEA 145 EUR
APOSOR110851-1G
5-Bromoindan-1-ol
5-Bromoindan-1-ol
Formel: C₉H₉BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD06808667
CAS-Nummer: 34598-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR