2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
Lieferant: Apollo Scientific
OR110855-1G
APOSOR110855-1GEA
116
EUR
APOSOR110855-1G
2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
Formel:
C₁₁H₁₃NO₄S Molecular Weight: 255,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001215 CAS-Nummer: 400878-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...