3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid

Lieferant: Apollo Scientific

OR110856-1G
APOSOR110856-1GEA 116 EUR
APOSOR110856-1G
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
Formel: C₁₁H₁₃NO₄S
Molecular Weight: 255,29 g/mol
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659598
CAS-Nummer: 763073-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR