3-(1,1-Dioxidothiomorpholin-4-yl)propanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR110858-1G
APOSOR110858-1GEA
116
EUR
APOSOR110858-1G
3-(1,1-Dioxidothiomorpholin-4-yl)propanoic acid
3-(1,1-Dioxidothiomorpholin-4-yl)propanoic acid
Formel:
C₇H₁₃NO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787455 CAS-Nummer: 849928-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...