2-(1,1-Dioxidothiomorpholin-4-yl)-3-methylbutanoic acid

Lieferant: Apollo Scientific

OR110859-1G
APOSOR110859-1GEA 116 EUR
APOSOR110859-1G
2-(1,1-Dioxidothiomorpholin-4-yl)-3-methylbutanoic acid
2-(1,1-Dioxidothiomorpholin-4-yl)-3-methylbutanoic acid
Formel: C₉H₁₇NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD02081981
CAS-Nummer: 477858-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR