2-(1,1-Dioxidothiomorpholin-4-yl)-3,3-dimethylbutanoic acid

Lieferant: Apollo Scientific

OR110860-1G
APOSOR110860-1GEA 232 EUR
APOSOR110860-1G
2-(1,1-Dioxidothiomorpholin-4-yl)-3,3-dimethylbutanoic acid
2-(1,1-Dioxidothiomorpholin-4-yl)-3,3-dimethylbutanoic acid
Formel: C₁₀H₁₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD01316045
CAS-Nummer: 338963-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR