6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine

Lieferant: Apollo Scientific

Synonyme: 6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-ylamine

OR110862-1G
APOSOR110862-1GEA 248 EUR
APOSOR110862-1G
6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine
6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine
Formel: C₆H₈N₂OS
Lagertemperatur: Kühlschrank
MDL: MFCD08690160
CAS-Nummer: 259810-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR