Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110864-250MG
APOSOR110864-250MGEA 244 EUR
APOSOR110864-250MG
Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Formel: C₁₀H₉BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22551971
CAS-Nummer: 1251033-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR