Ethyl 3-bromo-1H-indazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110865-1G
APOSOR110865-1GEA 368 EUR
APOSOR110865-1G
Ethyl 3-bromo-1H-indazole-5-carboxylate
Ethyl 3-bromo-1H-indazole-5-carboxylate
Formel: C₁₀H₉BrN₂O₂
Molecular Weight: 269.095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21607224
CAS-Nummer: 192945-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR