Ethyl 2-(methylthio)-4-phenylpyrimidine-5-carboxylate

Lieferant: Apollo Scientific

OR110866-1G OR110866-5G
APOSOR110866-1GEA 186 EUR
APOSOR110866-1G APOSOR110866-5G
Ethyl 2-(methylthio)-4-phenylpyrimidine-5-carboxylate
Ethyl 2-(methylthio)-4-phenylpyrimidine-5-carboxylate
Formel: C₁₄H₁₄N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD14897193
CAS-Nummer: 77995-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR