(4-Hydroxy-3-methoxybenzylidene)malononitrile

Lieferant: Apollo Scientific

OR110867-1G
APOSOR110867-1GEA 198 EUR
APOSOR110867-1G
(4-Hydroxy-3-methoxybenzylidene)malononitrile
(4-Hydroxy-3-methoxybenzylidene)malononitrile
Formel: C₁₁H₈N₂O₂
Molecular Weight: 200,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00137632
CAS-Nummer: 3696-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR