5-Methoxy-2-methyl-1,3-benzoxazole

Lieferant: Apollo Scientific

OR110870-1G
APOSOR110870-1GEA 0 EUR
APOSOR110870-1G
5-Methoxy-2-methyl-1,3-benzoxazole
5-Methoxy-2-methyl-1,3-benzoxazole
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128831
CAS-Nummer: 5676-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR