Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Lieferant: Apollo Scientific
OR110871-1G
APOSOR110871-1GEA
232
EUR
APOSOR110871-1G
Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Formel:
C₁₁H₁₁NO₃ Molecular Weight: 205,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124610 CAS-Nummer: 96017-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...