Methyl 2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoate

Lieferant: Apollo Scientific

OR110872-1G
APOSOR110872-1GEA 212 EUR
APOSOR110872-1G
Methyl 2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoate
Methyl 2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoate
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976394
CAS-Nummer: 96017-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR