Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate

Lieferant: Apollo Scientific

OR110874-1G
APOSOR110874-1GEA 232 EUR
APOSOR110874-1G
Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate
Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976395
CAS-Nummer: 1268618-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR