4-Phenyl-2-pyridin-4-ylpyrimidine-5-carboxylic acid

Lieferant: Apollo Scientific

OR110888-1G
APOSOR110888-1GEA 371 EUR
APOSOR110888-1G
4-Phenyl-2-pyridin-4-ylpyrimidine-5-carboxylic acid
4-Phenyl-2-pyridin-4-ylpyrimidine-5-carboxylic acid
Formel: C₁₆H₁₁N₃O₂
Molecular Weight: 277,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976392
CAS-Nummer: 311340-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR