5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
Lieferant: Apollo Scientific
OR110889-1G
APOSOR110889-1GEA
371
EUR
APOSOR110889-1G
5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
Formel:
C₁₀H₁₁N₃S₂ Molecular Weight: 237,344 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11137540 CAS-Nummer: 100988-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...