Ethyl 5-chloro-6-hydroxynicotinate

Lieferant: Apollo Scientific
Achtung

OR110895-1G
APOSOR110895-1GEA 372 EUR
APOSOR110895-1G
Ethyl 5-chloro-6-hydroxynicotinate
Ethyl 5-chloro-6-hydroxynicotinate
Formel: C₈H₈ClNO₃
Molecular Weight: 201,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11973933
CAS-Nummer: 58236-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR