Methyl 3-nitroimidazo[1,2-a]pyridine-7-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110896-1G
APOSOR110896-1GEA 383 EUR
APOSOR110896-1G
Methyl 3-nitroimidazo[1,2-a]pyridine-7-carboxylate
Methyl 3-nitroimidazo[1,2-a]pyridine-7-carboxylate
Formel: C₉H₇N₃O₄
Molecular Weight: 221,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995618
CAS-Nummer: 1823865-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR