5-Bromo-2-(1,3-oxazol-5-yl)pyridine
Lieferant: Apollo Scientific
Synonyme:
5-(5-Bromopyridin-2-yl)oxazole
OR110897-1G
APOSOR110897-1GEA
0
EUR
APOSOR110897-1G
5-Bromo-2-(1,3-oxazol-5-yl)pyridine
5-Bromo-2-(1,3-oxazol-5-yl)pyridine
Formel:
C₈H₅BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD27950166 CAS-Nummer: 380380-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...