tert-Butyl (5-nitro-1H-indol-1-yl)acetate

Lieferant: Apollo Scientific

OR110901-1G
APOSOR110901-1GEA 0 EUR
APOSOR110901-1G
tert-Butyl (5-nitro-1H-indol-1-yl)acetate
tert-Butyl (5-nitro-1H-indol-1-yl)acetate
Formel: C₁₄H₁₆N₂O₄
Molecular Weight: 276,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995627
CAS-Nummer: 227302-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR