4-Chloro-2-(1H-imidazol-1-yl)pyrimidine

Lieferant: Apollo Scientific

OR110904-250MG
APOSOR110904-250MGEA 473 EUR
APOSOR110904-250MG
4-Chloro-2-(1H-imidazol-1-yl)pyrimidine
4-Chloro-2-(1H-imidazol-1-yl)pyrimidine
Formel: C₇H₅ClN₄
Molecular Weight: 180.594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09831925
CAS-Nummer: 114834-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR