Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110907-1G OR110907-100MG OR110907-250MG
APOSOR110907-1GEA 290 EUR
APOSOR110907-1G APOSOR110907-100MG APOSOR110907-250MG
Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate
Ethyl 8-chloroimidazo[1,2-a]pyridine-6-carboxylate
Formel: C₁₀H₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27995635
CAS-Nummer: 1823330-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR