3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole

Lieferant: Apollo Scientific

OR110912-1G
APOSOR110912-1GEA 433 EUR
APOSOR110912-1G
3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole
3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole
Formel: C₁₁H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00112206
CAS-Nummer: 852690-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR