3-(5-Bromo-1-benzofuran-2-yl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR110917-1G
APOSOR110917-1GEA
346
EUR
APOSOR110917-1G
3-(5-Bromo-1-benzofuran-2-yl)-1H-pyrazole
3-(5-Bromo-1-benzofuran-2-yl)-1H-pyrazole
Formel:
C₁₁H₇BrN₂O Molecular Weight: 263,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00120853 CAS-Nummer: 666822-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...