Methyl 4-[3-(dimethylamino)prop-2-enoyl]benzoate

Lieferant: Apollo Scientific

OR110918-1G
APOSOR110918-1GEA 149 EUR
APOSOR110918-1G
Methyl 4-[3-(dimethylamino)prop-2-enoyl]benzoate
Methyl 4-[3-(dimethylamino)prop-2-enoyl]benzoate
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09908108
CAS-Nummer: 114431-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR