Methyl 6,7-dihydro-4H-thieno[3,2-c]pyran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110919-1G
APOSOR110919-1GEA 371 EUR
APOSOR110919-1G
Methyl 6,7-dihydro-4H-thieno[3,2-c]pyran-2-carboxylate
Methyl 6,7-dihydro-4H-thieno[3,2-c]pyran-2-carboxylate
Formel: C₉H₁₀O₃S
Molecular Weight: 198,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995615
CAS-Nummer: 1823277-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR