6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
Lieferant: Apollo Scientific
OR110921-1G
APOSOR110921-1GEA
174
EUR
APOSOR110921-1G
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
Formel:
C₁₀H₁₂N₂OS Molecular Weight: 208,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570181 CAS-Nummer: 215928-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...