1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6,7-Tetrahydropyrano[4,3-c]pyrazole

OR110924-1G
APOSOR110924-1GEA 386 EUR
APOSOR110924-1G
1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole
1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole
Formel: C₆H₈N₂O
Molecular Weight: 124.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18250670
CAS-Nummer: 258353-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR