6,7-Dihydro-4H-thieno[3,2-c]pyran-2-ylmethanol

Lieferant: Apollo Scientific

OR110925-1G
APOSOR110925-1GEA 395 EUR
APOSOR110925-1G
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-ylmethanol
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-ylmethanol
Formel: C₈H₁₀O₂S
Molecular Weight: 170,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169813
CAS-Nummer: 1265897-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR