6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR110926-1G
APOSOR110926-1GEA 551 EUR
APOSOR110926-1G
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carbaldehyde
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carbaldehyde
Formel: C₈H₈O₂S
Molecular Weight: 168,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18433145
CAS-Nummer: 623564-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR