Ethyl 3-morpholin-4-ylquinoxaline-2-carboxylate

Lieferant: Apollo Scientific

OR110928-1G
APOSOR110928-1GEA 296 EUR
APOSOR110928-1G
Ethyl 3-morpholin-4-ylquinoxaline-2-carboxylate
Ethyl 3-morpholin-4-ylquinoxaline-2-carboxylate
Formel: C₁₅H₁₇N₃O₃
Molecular Weight: 287,314 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995620
CAS-Nummer: 108222-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR