3-Morpholin-4-ylquinoxaline-2-carboxylic acid

Lieferant: Apollo Scientific

OR110930-1G
APOSOR110930-1GEA 493 EUR
APOSOR110930-1G
3-Morpholin-4-ylquinoxaline-2-carboxylic acid
3-Morpholin-4-ylquinoxaline-2-carboxylic acid
Formel: C₁₃H₁₃N₃O₃
Molecular Weight: 259,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15204303
CAS-Nummer: 1211503-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR