tert-Butyl 4-[3-(ethoxycarbonyl)quinoxalin-2-yl]homopiperazine-1-carboxylate
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 4-[3-(ethoxycarbonyl)quinoxalin-2-yl]-1,4-diazepane-1-carboxylate
OR110931-1G
APOSOR110931-1GEA
445
EUR
APOSOR110931-1G
tert-Butyl 4-[3-(ethoxycarbonyl)quinoxalin-2-yl]homopiperazine-1-carboxylate
tert-Butyl 4-[3-(ethoxycarbonyl)quinoxalin-2-yl]homopiperazine-1-carboxylate
Formel:
C₂₁H₂₈N₄O₄ Molecular Weight: 400,471 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27995631 CAS-Nummer: 1858255-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...