2-(2-(2-(3-Pyridinyl)-1,3-thiazol-4-yl)ethyl)-1H-isoindole-1,3(2H)-dione

Lieferant: Apollo Scientific

OR110934-1G
APOSOR110934-1GEA 371 EUR
APOSOR110934-1G
2-(2-(2-(3-Pyridinyl)-1,3-thiazol-4-yl)ethyl)-1H-isoindole-1,3(2H)-dione
2-(2-(2-(3-Pyridinyl)-1,3-thiazol-4-yl)ethyl)-1H-isoindole-1,3(2H)-dione
Formel: C₁₈H₁₃N₃O₂S
Molecular Weight: 335,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124546
CAS-Nummer: 860785-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR