3-Bromo-5-methylimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR110936-1G
APOSOR110936-1GEA 85.7 EUR
APOSOR110936-1G
3-Bromo-5-methylimidazo[1,2-a]pyridine
3-Bromo-5-methylimidazo[1,2-a]pyridine
Formel: C₈H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19441944
CAS-Nummer: 5857-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR