2-Methyl-6-nitroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR110937-1G
APOSOR110937-1GEA 232 EUR
APOSOR110937-1G
2-Methyl-6-nitroimidazo[1,2-a]pyridine
2-Methyl-6-nitroimidazo[1,2-a]pyridine
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05863412
CAS-Nummer: 13212-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR