2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
Lieferant: Apollo Scientific
OR110940-1G
OR110940-250MG
OR110940-500MG
APOSOR110940-1GEA
43.5
EUR
APOSOR110940-1G
APOSOR110940-250MG
APOSOR110940-500MG
2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
Formel:
C₇H₅ClN₄ Molecular Weight: 180.6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09756941 CAS-Nummer: 114834-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...