Ethyl-(4-iodphenyl)acetat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-(4-iodophenyl)acetate
OR110943-1G
OR110943-25G
OR110943-5G
APOSOR110943-1GEA
21.9
EUR
APOSOR110943-1G
APOSOR110943-25G
APOSOR110943-5G
Ethyl-(4-iodphenyl)acetat
Ethyl-(4-iodphenyl)acetat
Formel:
C₁₀H₁₁IO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779004 CAS-Nummer: 15250-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...